SY3H3D6805DA Synthonix Corporation

3-(3-Methoxyphenyl)-1-propene

  • MDL number No MDL found
  • Cas Number : 24743-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52782.html

Safety Information

MSDS https://synthonix.com/52782.html
Availability details for
Quantity
  • Availability
  • Add to Cart