SY3H3D6805DB Synthonix Corporation

3'-Methoxypropiophenone

  • MDL number No MDL found
  • Cas Number : 37951-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52788.html

Safety Information

MSDS https://synthonix.com/52788.html
Availability details for
Quantity
  • Availability
  • Add to Cart