SY3H3D6805DD Synthonix Corporation

Methyl 2,3-dimethylbenzoate

  • MDL number No MDL found
  • Cas Number : 15012-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52791.html

Safety Information

MSDS https://synthonix.com/52791.html
Availability details for
Quantity
  • Availability
  • Add to Cart