SY3H3D6805EB Synthonix Corporation

1-(3-Methoxy-5-methylphenyl)ethanone

  • MDL number No MDL found
  • Cas Number : 43113-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52814.html

Safety Information

MSDS https://synthonix.com/52814.html
Availability details for
Quantity
  • Availability
  • Add to Cart