SY3H3D6805EC Synthonix Corporation

5-Methoxychroman-4-ol

  • MDL number MFCD27921584
  • Cas Number : 1616526-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52815.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52815.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart