SY3H3D6805ED Synthonix Corporation

Methyl 2-Methoxyphenylacetate

  • MDL number MFCD02093479
  • Cas Number : 27798-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52820.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52820.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart