SY3H3D6805F5 Synthonix Corporation

Methyl 4-methoxy-2-methylbenzoate

  • MDL number No MDL found
  • Cas Number : 35598-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/52833.html

Safety Information

MSDS https://synthonix.com/52833.html
Availability details for
Quantity
  • Availability
  • Add to Cart