SY3H3D6805F8 Synthonix Corporation

1-(2-Methoxyphenoxy)-2,3-epoxypropane

  • MDL number MFCD00190158
  • Cas Number : 2210-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Lump / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52842.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52842.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart