SY3H3D6805F9 Synthonix Corporation

Methyl 4-hydroxy-2,6-dimethylbenzoate

  • MDL number No MDL found
  • Cas Number : 83194-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52843.html

Safety Information

MSDS https://synthonix.com/52843.html
Availability details for
Quantity
  • Availability
  • Add to Cart