SY3H3D6805FC Synthonix Corporation

Methyl 2-(4-hydroxy-3-methylphenyl)acetate

  • MDL number No MDL found
  • Cas Number : 64360-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/52848.html

Safety Information

MSDS https://synthonix.com/52848.html
Availability details for
Quantity
  • Availability
  • Add to Cart