SY3H3D680602 Synthonix Corporation

METHYL 2-HYDROXY-3-(3-HYDROXYPHENYL)PROPANOATE

  • MDL number No MDL found
  • Cas Number : 1508892-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52860.html

Safety Information

MSDS https://synthonix.com/52860.html
Availability details for
Quantity
  • Availability
  • Add to Cart