SY3H3D680603 Synthonix Corporation

Methyl 2-methoxy-5-(methylsulfonyl)benzoate

  • MDL number No MDL found
  • Cas Number : 63484-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52862.html

Safety Information

MSDS https://synthonix.com/52862.html
Availability details for
Quantity
  • Availability
  • Add to Cart