SY3H3D680604 Synthonix Corporation

3-Methyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD09896588
  • Cas Number : 20668-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Colorless to light-yellow to yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52877.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52877.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart