SY3H3D680616 Synthonix Corporation

Methyl 4-amino-2,6-dimethylbenzoate

  • MDL number No MDL found
  • Cas Number : 79909-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52959.html

Safety Information

MSDS https://synthonix.com/52959.html
Availability details for
Quantity
  • Availability
  • Add to Cart