SY3H3D68061A Synthonix Corporation

Methyl 3-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-2-carboxylate

  • Cas Number : 119205-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52973.html

Safety Information

MSDS https://synthonix.com/52973.html
Availability details for
Quantity
  • Availability
  • Add to Cart