SY3H3D68061D Synthonix Corporation

Methyl 2-amino-4,6-dimethoxybenzoate

  • Cas Number : 379228-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52992.html

Safety Information

MSDS https://synthonix.com/52992.html
Availability details for
Quantity
  • Availability
  • Add to Cart