SY3H3D68061E Synthonix Corporation

5-(Morpholinomethyl)furan-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 26095-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52993.html

Safety Information

MSDS https://synthonix.com/52993.html
Availability details for
Quantity
  • Availability
  • Add to Cart