SY3H3D680626 Synthonix Corporation

(4-Morpholinophenyl)boronic acid

  • MDL number No MDL found
  • Cas Number : 186498-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53077.html

Safety Information

MSDS https://synthonix.com/53077.html
Availability details for
Quantity
  • Availability
  • Add to Cart