SY3H3D680629 Synthonix Corporation

1-Methoxy-2,3,5-trimethylbenzene

  • MDL number MFCD00142783
  • Cas Number : 20469-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53084.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53084.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart