SY3H3D68062F Synthonix Corporation

N-(4-Methylbenzyl)ethanamine

  • MDL number No MDL found
  • Cas Number : 39099-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53114.html

Safety Information

MSDS https://synthonix.com/53114.html
Availability details for
Quantity
  • Availability
  • Add to Cart