SY3H3D680631 Synthonix Corporation

(S)-2-Methyl-1-phenylpropan-1-amine

  • MDL number No MDL found
  • Cas Number : 68906-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53117.html

Safety Information

MSDS https://synthonix.com/53117.html
Availability details for
Quantity
  • Availability
  • Add to Cart