SY3H3D680632 Synthonix Corporation

1-(3-Methoxypyridin-2-yl)piperazine

  • MDL number No MDL found
  • Cas Number : 80827-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/53126.html

Safety Information

MSDS https://synthonix.com/53126.html
Availability details for
Quantity
  • Availability
  • Add to Cart