SY3H3D680633 Synthonix Corporation

N-(3-Methoxybenzyl)ethanamine

  • Cas Number : 140715-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53138.html

Safety Information

MSDS https://synthonix.com/53138.html
Availability details for
Quantity
  • Availability
  • Add to Cart