SY3H3D680639 Synthonix Corporation

(6-(4-Methylpiperazin-1-yl)pyridin-3-yl)boronic acid

  • MDL number MFCD09907981
  • Cas Number : 936353-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53199.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53199.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart