SY3H3D68063A Synthonix Corporation

2-(4-Methylpiperazin-1-yl)pyridin-3-amine

  • MDL number No MDL found
  • Cas Number : 5028-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53201.html

Safety Information

MSDS https://synthonix.com/53201.html
Availability details for
Quantity
  • Availability
  • Add to Cart