SY3H3D68063E Synthonix Corporation

trans-4-Morpholinocyclohexanol

  • Cas Number : 1228947-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53287.html

Safety Information

MSDS https://synthonix.com/53287.html
Availability details for
Quantity
  • Availability
  • Add to Cart