SY3H3D68063F Synthonix Corporation

Methyl 3-((tert-butoxycarbonyl)(methyl)amino)propanoate

  • MDL number No MDL found
  • Cas Number : 119740-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53303.html

Safety Information

MSDS https://synthonix.com/53303.html
Availability details for
Quantity
  • Availability
  • Add to Cart