SY3H3D680640 Synthonix Corporation

4-(Morpholinomethyl)piperidin-4-ol

  • Cas Number : 885654-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53321.html

Safety Information

MSDS https://synthonix.com/53321.html
Availability details for
Quantity
  • Availability
  • Add to Cart