SY3H3D680641 Synthonix Corporation

4-Methylnonanoic acid

  • Cas Number : 45019-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53337.html

Safety Information

MSDS https://synthonix.com/53337.html
Availability details for
Quantity
  • Availability
  • Add to Cart