SY3H3D680644 Synthonix Corporation

Methyl 2-chloroquinoxaline-6-carboxylate

  • Cas Number : 2090584-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53528.html

Safety Information

MSDS https://synthonix.com/53528.html
Availability details for
Quantity
  • Availability
  • Add to Cart