SY3H3D680647 Synthonix Corporation

Methyl 2,3-dioxoindoline-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 153072-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53584.html

Safety Information

MSDS https://synthonix.com/53584.html
Availability details for
Quantity
  • Availability
  • Add to Cart