SY3H3D680649 Synthonix Corporation

Methyl 7-nitrobenzo[b]thiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 34084-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53588.html

Safety Information

MSDS https://synthonix.com/53588.html
Availability details for
Quantity
  • Availability
  • Add to Cart