SY3H3D68064B Synthonix Corporation

Methyl 6-chlorobenzo[b]thiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 104795-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53597.html

Safety Information

MSDS https://synthonix.com/53597.html
Availability details for
Quantity
  • Availability
  • Add to Cart