SY3H3D68064C Synthonix Corporation

Methyl 4-ethynyl-3-fluorobenzoate

  • MDL number No MDL found
  • Cas Number : 1459216-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/53598.html

Safety Information

MSDS https://synthonix.com/53598.html
Availability details for
Quantity
  • Availability
  • Add to Cart