SY3H3D68064D Synthonix Corporation

Methyl 4-iodobenzo[b]thiophene-2-carboxylate

  • Cas Number : 146137-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53603.html

Safety Information

MSDS https://synthonix.com/53603.html
Availability details for
Quantity
  • Availability
  • Add to Cart