SY3H3D68064E Synthonix Corporation

Methyl 7-chloro-3-hydroxybenzo[b]thiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 33851-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53608.html

Safety Information

MSDS https://synthonix.com/53608.html
Availability details for
Quantity
  • Availability
  • Add to Cart