SY3H3D68064F Synthonix Corporation

3-Methyl-1H-indole-2-carbonitrile

  • MDL number No MDL found
  • Cas Number : 13006-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53633.html

Safety Information

MSDS https://synthonix.com/53633.html
Availability details for
Quantity
  • Availability
  • Add to Cart