SY3H3D68065A Synthonix Corporation

4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoic acid

  • MDL number No MDL found
  • Cas Number : 95124-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53674.html

Safety Information

MSDS https://synthonix.com/53674.html
Availability details for
Quantity
  • Availability
  • Add to Cart