SY3H3D68065B Synthonix Corporation

Methyl 2-oxo-1,2-dihydroquinazoline-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 1036755-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/53676.html

Safety Information

MSDS https://synthonix.com/53676.html
Availability details for
Quantity
  • Availability
  • Add to Cart