SY3H3D68065D Synthonix Corporation

Methyl 5-formylpyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 936637-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53678.html

Safety Information

MSDS https://synthonix.com/53678.html
Availability details for
Quantity
  • Availability
  • Add to Cart