SY3H3D68065E Synthonix Corporation

4-Methoxy-6-nitroquinolin-2(1H)-one

  • MDL number No MDL found
  • Cas Number : 934687-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53680.html

Safety Information

MSDS https://synthonix.com/53680.html
Availability details for
Quantity
  • Availability
  • Add to Cart