SY3H3D680660 Synthonix Corporation

1-Methyl-5-(trifluoromethyl)-1H-indole

  • Cas Number : 1264670-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53738.html

Safety Information

MSDS https://synthonix.com/53738.html
Availability details for
Quantity
  • Availability
  • Add to Cart