SY3H3D680C6D Synthonix Corporation

4-Methyl-1,2,3,4-tetrahydroquinoline hydrochloride

  • MDL number No MDL found
  • Cas Number : 74459-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/54141.html

Safety Information

MSDS https://synthonix.com/54141.html
Availability details for
Quantity
  • Availability
  • Add to Cart