SY3H3D680C6E Synthonix Corporation

4-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number No MDL found
  • Cas Number : 111661-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54144.html

Safety Information

MSDS https://synthonix.com/54144.html
Availability details for
Quantity
  • Availability
  • Add to Cart