SY3H3D680C70 Synthonix Corporation

Methyl 6-fluoro-1,2,3,4-tetrahydroisoquinoline-1-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 2007908-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55451.html

Safety Information

MSDS https://synthonix.com/55451.html
Availability details for
Quantity
  • Availability
  • Add to Cart