SY3H3D680C71 Synthonix Corporation

5-Methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1989671-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55486.html

Safety Information

MSDS https://synthonix.com/55486.html
Availability details for
Quantity
  • Availability
  • Add to Cart