SY3H3D680C72 Synthonix Corporation

7-Methyl-2,3,4,5-tetrahydro-1H-benzo[b]azepin-5-amine dihydrochloride

  • Cas Number : 1416354-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55551.html

Safety Information

MSDS https://synthonix.com/55551.html
Availability details for
Quantity
  • Availability
  • Add to Cart