SY3H3D680C74 Synthonix Corporation

Methyl 6-chloro-1H-indole-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 921194-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53760.html

Safety Information

MSDS https://synthonix.com/53760.html
Availability details for
Quantity
  • Availability
  • Add to Cart