SY3H3D680C75 Synthonix Corporation

2-Methyl-5-(trifluoromethoxy)-1H-indole

  • Cas Number : 900182-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53791.html

Safety Information

MSDS https://synthonix.com/53791.html
Availability details for
Quantity
  • Availability
  • Add to Cart