SY3H3D680C78 Synthonix Corporation

3-Methylbenzofuran-2-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 1199-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53796.html

Safety Information

MSDS https://synthonix.com/53796.html
Availability details for
Quantity
  • Availability
  • Add to Cart